Läs mer
1-Amino-1-cyclopentanecarboxylic Acid Methyl Ester, TRC
Produktkod. 30317779
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30317779 1 g
1 options
Produktkod. 30317779 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6035451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Amino-1-cyclopentanecarboxylic Acid Methyl Ester
CAS 78388-61-1
Molekylformel C7 H13 N O2
Kvantitet 1 g
Synonym Cyclopentanecarboxylic acid, 1-amino-, methyl ester, 1-Aminocyclopentanecarboxylic acid methyl ester, Cycloleucine methyl ester, Methyl 1-amino-1-cyclopentanecarboxylate, Methyl 1-aminocyclopentanecarboxylate, NSC 26981, 1-Amino-1-cyclopentanecarboxylic acid methyl ester
InChI formel InChI=1S/C7H13NO2/c1-10-6(9)7(8)4-2-3-5-7/h2-5,8H2,1H3
LEDER COC(=O)C1(N)CCCC1
IUPAC-namn methyl 1-aminocyclopentane-1-carboxylate
Molekylvikt (g/mol) 143.18
Formel vikt 143.0946