Läs mer
1-Amino-2,4-dibromo-anthraquinone, TRC
Produktkod. 30313897
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30313897 100 mg
30323301 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30313897 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA604460100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Amino-2,4-dibromo-anthraquinone
CAS 81-49-2
Molekylformel C14H7Br2NO2
Kvantitet 100 mg
Synonym NSC 3529, 1-Amino-2,4-dibromo-9,10-anthracenedione, 1-Amino-2,4-dibromo-9,10-anthraquinone, 1-Amino-2,4-dibromoanthraquinone, 2,4-Dibromo-1-anthraquinonylamine
InChI formel InChI=1S/C14H7Br2NO2/c15-8-5-9(16)12(17)11-10(8)13(18)6-3-1-2-4-7(6)14(11)19/h1-5H,17H2
LEDER C1=CC2=C(C=C1)C(=O)C3=C(C(=CC(=C3C2=O)Br)Br)N
IUPAC-namn 1-amino-2,4-dibromoanthracene-9,10-dione
Molekylvikt (g/mol) 381.02
Formel vikt 378.8844