Läs mer
1-Amino-2-naphthol hydrochloride, 97%, Thermo Scientific™
Produktkod. 11455957
Change view
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11455957 25 g 25g
11435957 1 g 1g
11445957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11455957 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B24492.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1198-27-2
Molekylformel C10H10ClNO
Molekylvikt (g/mol) 195.646
MDL-nummer MFCD00012561
InChI-nyckel DEKREBCFNLUULC-UHFFFAOYSA-N
Synonym 1-amino-2-naphthol hydrochloride, 1-aminonaphthalen-2-ol hydrochloride, 1-amino-2-naphthol hcl, unii-xm2lzu54ow, 2-naphthalenol, 1-amino-, hydrochloride 1:1, 2-naphthalenol, 1-amino-, hydrochloride, xm2lzu54ow, 1-aminonaphthalen-2-ol, chloride, timtec-bb sbb000347, acmc-209a4b
PubChem CID 518469
IUPAC-namn 1-aminonaphthalen-2-ol;hydrochloride
LEDER C1=CC=C2C(=C1)C=CC(=C2N)O.Cl
Specifikationer
Kemiskt namn eller material 1-Amino-2-naphthol hydrochloride
Smältpunkt ∼276°C (decomposition)
Kvantitet 25 g
Beilstein 3699817
Känslighet Light sensitive
Formel vikt 195.65
Procent renhet 97%