Läs mer
1-Amino-2-naphthol Hydrochloride, TRC
Produktkod. 30324159
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30324159 500 mg
30314900 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30324159 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA618723500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Amino-2-naphthol Hydrochloride
CAS 1198-27-2
Molekylformel C10H9NO . HCl
Kvantitet 500 mg
InChI formel InChI=1S/C10H9NO.ClH/c11-10-8-4-2-1-3-7(8)5-6-9(10)12;/h1-6,12H,11H2;1H
LEDER Cl.Nc1c(O)ccc2ccccc12
IUPAC-namn 1-aminonaphthalen-2-ol;hydrochloride
Molekylvikt (g/mol) 195.64
Formel vikt 195.0451