missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Amino-4-(2-hydroxyethyl)piperazine, 96%

Brand:  Alfa Aesar™ H55134.06

Additional Details : CAS Number : 3973-70-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15481288

  • 7819.25 SEK / 5g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

1-Amino-4-(2-hydroxyethyl)piperazine
145.2
4-Amino-1-piperazineethanol
3973-70-4
MFCD00006155

Specifications

1-Amino-4-(2-hydroxyethyl)piperazine
Powder
105° to 107°C
MFCD00006155
5g
3973-70-4
96%
145.2
White to pale brown
4-Amino-1-piperazineethanol
Videos
SDS