Learn More
1-Amino-4-cyclopentylpiperazine, 97%, Thermo Scientific Chemicals
Catalog Number p-7036524
Click to view available options
Quantity:
250 mg
1 g
5 g
61379-64-4
C9H19N3
169.272
MFCD00190598
QYHRIASMJNLWHJ-UHFFFAOYSA-N
1512485
4-cyclopentylpiperazin-1-amine
C1CCC(C1)N2CCN(CC2)N
61379-64-4
C9H19N3
169.272
MFCD00190598
QYHRIASMJNLWHJ-UHFFFAOYSA-N
1512485
4-cyclopentylpiperazin-1-amine
C1CCC(C1)N2CCN(CC2)N
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 1-amino-4-cyclopentylpiperazine, 1-amino-4-cyclopentyl piperazine, 4-cyclopentyl-1-piperazinamine, pubchem8581, 4-cyclopentylpiperazinylamine, 1-piperazinamine,4-cyclopentyl, 1-piperazinamine, 4-cyclopentyl, 4-cyclopentyl-piperazin-1-ylamine, 1-n-cyclopentyl piperazine-4-amine, 1-piperazinamine, 4-cyclopentyl-9ci
Solubility Information Slightly soluble in water.
Formula Weight 169.27
Percent Purity 97%
Chemical Name or Material 1-Amino-4-cyclopentylpiperazine