Läs mer
1-Aminoacetone Hydrochloride, TRC
Produktkod. 30326072
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30326072 25 mg
1 options
Produktkod. 30326072 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA58006025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Aminoacetone, Hydrochloride
CAS 7737-17-9
Molekylformel C3 H7 N O . Cl H
Kvantitet 25 mg
Synonym 2-Propanone, amino-, hydrochloride (6CI), 1-Amino-2-propanone hydrochloride, 1-Aminoacetone Hydrochloride, 1-Aminopropan-2-one hydrochloride salt, 2-Oxopropylammonium chloride, Aminoacetone hydrochloride
InChI formel InChI=1S/C3H7NO.ClH/c1-3(5)2-4;/h2,4H2,1H3;1H
LEDER Cl.CC(=O)CN
IUPAC-namn 1-aminopropan-2-one;hydrochloride
Molekylvikt (g/mol) 109.55
Formel vikt 109.0294