missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopropane-2,2,3,3-d4-carboxylic Acid, TRC

Product Code. 30311718
Change view
Click to view available options
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30311718 1 mg
30309916 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311718 Supplier Toronto Research Chemicals Supplier No. TRCA6035511MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-Aminocyclopropane-1-carboxylic Acid-2,2,3,3-D4
CAS 84392-07-4
Molekylformel C4 D4 H3 N O2
Kvantitet 1 mg
Synonym Cyclopropane-2,2,3,3-d4-carboxylic acid, 1-amino- (9CI), 1-Aminocyclopropane-1-carboxylic Acid-2,2,3,3-D4, 1-Aminocyclopropane-2,2,3,3-d4-carboxylic acid
InChI formel InChI=1S/C4H7NO2/c5-4(1-2-4)3(6)7/h1-2,5H2,(H,6,7)/i1D2,2D2
LEDER [2H]C1([2H])C([2H])([2H])C1(N)C(=O)O
IUPAC-namn 1-amino-2,2,3,3-tetradeuteriocyclopropane-1-carboxylic acid
Molekylvikt (g/mol) 105.1285
Formel vikt 105.0728
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.