Läs mer
1-Aminopyrrolidin-2-one Hydrochloride, TRC
Produktkod. 30322707
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30322707 1 g
1 options
Produktkod. 30322707 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6288051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Aminopyrrolidin-2-one Hydrochloride
CAS 20386-22-5
Molekylformel C4H9ClN2O
Kvantitet 1 g
Synonym 1-Amino-2-pyrrolidinone Hydrochloride, 1-Amino-2-pyrrolidinone Monohydrochloride, 1-Aminopyrrolidin-2-one Hydrochloride, N-Amino-2-pyrrolidinone Hydrochloride
InChI formel InChI=1S/C4H8N2O.ClH/c5-6-3-1-2-4(6)7;/h1-3,5H2;1H
LEDER O=C1N(N)CCC1.[H]Cl
IUPAC-namn 1-aminopyrrolidin-2-one hydrogen chloride
Molekylvikt (g/mol) 136.58
Formel vikt 136.04