missing translation for 'onlineSavingsMsg'
Läs mer

1-Aminopyrrolidin-2-one Hydrochloride, TRC

Produktkod. 30325083
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30325083 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. View return policy
Product Code. 30325083 Supplier Toronto Research Chemicals Supplier No. TRCA62880550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-Aminopyrrolidin-2-one Hydrochloride
CAS 20386-22-5
Molekylformel C4H9ClN2O
Kvantitet 50 g
Synonym 1-Amino-2-pyrrolidinone Hydrochloride, 1-Amino-2-pyrrolidinone Monohydrochloride, 1-Aminopyrrolidin-2-one Hydrochloride, N-Amino-2-pyrrolidinone Hydrochloride
InChI formel InChI=1S/C4H8N2O.ClH/c5-6-3-1-2-4(6)7;/h1-3,5H2;1H
LEDER O=C1N(N)CCC1.[H]Cl
IUPAC-namn 1-aminopyrrolidin-2-one hydrogen chloride
Molekylvikt (g/mol) 136.58
Formel vikt 136.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.