Läs mer
1-Anilino-3-phenylimino-1-propene Hydrochloride, TRC
Produktkod. 30318142
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30318142 5 g
1 options
Produktkod. 30318142 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA4589205G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Anilino-3-phenylimino-1-propene Hydrochloride
CAS 50328-50-2
Molekylformel C15H14N2 . xHCl
Kvantitet 5 g
Synonym N-[3-(Phenylamino)-2-propenylidene]benzenamine Hydrochloride, N,N'-1-propen-1-yl-3-ylidene-aniline Hydrochloride
InChI formel InChI=1S/C15H14N2.ClH/c1-3-8-14(9-4-1)16-12-7-13-17-15-10-5-2-6-11-15;/h1-13,16H;1H/b12-7+,17-13+;
LEDER Cl.N(\C=C\C=N\c1ccccc1)c2ccccc2
IUPAC-namn N-((1E,3E)-3-(phenylimino)prop-1-en-1-yl)aniline hydrochloride
Molekylvikt (g/mol) 258.74