Läs mer
STEMCELL Technologies 1-Azakenpaullone
Beskrivning
- 1-Azakenpaullone is a potent and highly selective inhibitor of glycogen synthase kinase 3β (IC50 = 18 nM) and is highly preferential to GSK3β versus other cyclin-dependent kinases (CDKs) including CDK/cyclin B and CDK5/p25 (IC50 = 2 μM and 4.2 μM, respectively; Kunick et al.)
- In combination with ID-8 (Catalog #72502) and FK506 (Catalog #74152), supports human pluripotent stem cells in long-term propagation, maintaining pluripotency, and capacity to differentiate (Yasuda et al.)
Specifikationer
Specifikationer
| CAS | 676596-65-9 |
| Kvantitet | 5 mg |
| Molekylformel | C15H10BrN3O |
| Renhet | ≥98% |