Läs mer
1-Benzo[b]thien-4-yl-piperazine Hydrochloride, Mikromol
Produktkod. 30460853
Change view
Kvantitet:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460853 4 x 25 mg
30049841 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30460853 Leverantör Mikromol Leverantörsnummer MM3702.0100254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Benzo[b]thien-4-yl-piperazine Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Benzo[b]thien-4-yl-piperazine Hydrochloride
CAS 913614-18-3
Analyt- eller komponentnamn 1-Benzo[b]thien-4-yl-piperazine Hydrochloride
Frakt skick Room Temperature
Molekylformel C12 H14 N2 S . Cl H
Certifikat och efterlevnad ISO 17025
Synonym 1-Benzo[b]thien-4-yl-piperazine Hydrochloride, Piperazine, 1-benzo[b]thien-4-yl-, monohydrochloride (9CI)
InChI formel InChI=1S/C12H14N2S.ClH/c1-2-11(14-7-5-13-6-8-14)10-4-9-15-12(10)3-1;/h1-4,9,13H,5-8H2;1H
Lösningstyp Neat
LEDER Cl.C1CN(CCN1)c2cccc3sccc23
IUPAC-namn 1-(1-benzothiophen-4-yl)piperazine;hydrochloride
Molekylvikt (g/mol) 254.78
Kvantitet 4 x 25 mg
Formel vikt 254.0644 g/mol