Läs mer
1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride, Mikromol™
Beskrivning
1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride |
| CAS | 2731998-18-6 |
| Analyt- eller komponentnamn | 1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C14 H21 N3 O . 2 Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Azelastine Hydrochloride Imp. B (EP), Azelastine Imp. B (EP), Azelastine USP Related Compound B, Azelastine USP RC B, 1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride, Azelastine Hydrochloride Imp. B (EP) as Dihydrochloride, Azelastine Related Compound B as Dihydrochloride, Azelastine Hydrochloride Impurity B as Dihydrochloride, Azelastine Impurity B as Dihydrochloride |
| InChI formel | InChI=1S/C14H21N3O.2ClH/c1-17-10-5-8-13(9-11-17)15-16-14(18)12-6-3-2-4-7-12;;/h2-4,6-7,13,15H,5,8-11H2,1H3,(H,16,18);2*1H |
| LEDER | Cl.Cl.CN1CCCC(CC1)NNC(=O)c2ccccc2 |
| IUPAC-namn | N'-(1-methylazepan-4-yl)benzohydrazide;dihydrochloride |
| Visa mer |