missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-1,4,7,10-tetraazacyclododecane, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Used as ligands for catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as ligands for catalysts.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents.
Used as ligands for catalysts.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents.
Specifications
Melting Point | 84°C to 86°C |
Quantity | 250 mg |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 262.4 |
Chemical Name or Material | 1-Benzyl-1,4,7,10-tetraazacyclododecane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'