missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-cyano-4-hydroxypiperidine, 98%, Thermo Scientific™

Catalog Number p-65329
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6094-60-6
C13H16N2O
216.28
MFCD00023752
XQRLXUYZKZXSBN-UHFFFAOYSA-N
1-benzyl-4-cyano-4-hydroxypiperidine, maybridge1_006722, acmc-20aoxx, 1-benzyl-4-hydroxyisonipecotonitrile, 4-hydroxy-1-benzylpiperidine-4-carbonitrile, 1-benyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-hydroxy-4-piperidinecarbonitrile, 1-benzyl-4-hydroxy-piperidine-4-carbonitrile, 4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile
80189
1-benzyl-4-hydroxypiperidine-4-carbonitrile
OC1(CCN(CC2=CC=CC=C2)CC1)C#N
This item is not returnable. View return policy
6094-60-6
C13H16N2O
216.28
MFCD00023752
XQRLXUYZKZXSBN-UHFFFAOYSA-N
1-benzyl-4-cyano-4-hydroxypiperidine, maybridge1_006722, acmc-20aoxx, 1-benzyl-4-hydroxyisonipecotonitrile, 4-hydroxy-1-benzylpiperidine-4-carbonitrile, 1-benyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-hydroxy-4-piperidinecarbonitrile, 1-benzyl-4-hydroxy-piperidine-4-carbonitrile, 4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile
80189
1-benzyl-4-hydroxypiperidine-4-carbonitrile
OC1(CCN(CC2=CC=CC=C2)CC1)C#N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 95°C to 102°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 216.28
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-Benzyl-4-cyano-4-hydroxypiperidine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'