missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific™

Catalog Number p-7081109
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
4727-72-4
C12H17NO
191.274
MFCD00006503
BPPZXJZYCOETDA-UHFFFAOYSA-N
1-benzyl-4-hydroxypiperidine, 1-benzyl-4-piperidinol, n-benzyl-4-hydroxypiperidine, 1-benzyl-piperidin-4-ol, 4-piperidinol, 1-phenylmethyl, n-benzyl-4-hydroxy piperidine, n-benzyl-4-piperidinol, 1-benzyl-4-hydroxy piperidine, 1-phenylmethyl piperidin-4-ol, 1-phenylmethyl-4-piperidinol
78461
1-benzylpiperidin-4-ol
C1CN(CCC1O)CC2=CC=CC=C2
This item is not returnable. View return policy
4727-72-4
C12H17NO
191.274
MFCD00006503
BPPZXJZYCOETDA-UHFFFAOYSA-N
1-benzyl-4-hydroxypiperidine, 1-benzyl-4-piperidinol, n-benzyl-4-hydroxypiperidine, 1-benzyl-piperidin-4-ol, 4-piperidinol, 1-phenylmethyl, n-benzyl-4-hydroxy piperidine, n-benzyl-4-piperidinol, 1-benzyl-4-hydroxy piperidine, 1-phenylmethyl piperidin-4-ol, 1-phenylmethyl-4-piperidinol
78461
1-benzylpiperidin-4-ol
C1CN(CCC1O)CC2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 60°C to 64°C
Boiling Point 121°C to 123°C (0.7 mmHg)
Quantity 5 g
Beilstein 146118
Sensitivity Hygroscopic
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'