missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-(methylamino)piperidine, 98%, Thermo Scientific Chemicals

Catalog Number p-7030070
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
7006-50-0
C13H20N2
204.32
MFCD03931054
RGEQSTMITLEXKD-UHFFFAOYSA-N
1-benzyl-piperidin-4-yl-methyl-amine, 4-methylamino-1-benzylpiperidine, 1-benzyl-4-methylamino piperidine, 4-piperidinamine, n-methyl-1-phenylmethyl, 1-benzyl-n-methyl-piperidin-4-amine, pubchem7928, benzylmethylpiperidinamine, 1-benzyl-4-methylaminopiperidine, 4-methylamino-1-benzyl piperidine
4136128
1-benzyl-N-methylpiperidin-4-amine
CNC1CCN(CC2=CC=CC=C2)CC1
This item is not returnable. View return policy
7006-50-0
C13H20N2
204.32
MFCD03931054
RGEQSTMITLEXKD-UHFFFAOYSA-N
1-benzyl-piperidin-4-yl-methyl-amine, 4-methylamino-1-benzylpiperidine, 1-benzyl-4-methylamino piperidine, 4-piperidinamine, n-methyl-1-phenylmethyl, 1-benzyl-n-methyl-piperidin-4-amine, pubchem7928, benzylmethylpiperidinamine, 1-benzyl-4-methylaminopiperidine, 4-methylamino-1-benzyl piperidine
4136128
1-benzyl-N-methylpiperidin-4-amine
CNC1CCN(CC2=CC=CC=C2)CC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 204.31
Percent Purity 98%
Chemical Name or Material 1-Benzyl-4-(methylamino)piperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'