Läs mer
1-Benzyl-5-(dicyclohexylcarbodiimido)glutarate, TRC
Produktkod. 30346919
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30346919 1 g
1 options
Product Code. 30346919 Supplier Toronto Research Chemicals Supplier No. TRCB2756401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 1-Benzyl-5-(dicyclohexylcarbodiimido)glutarate
CAS 887352-83-2
Molekylformel C25 H36 N2 O4
Kvantitet 1 g
InChI formel InChI=1S/C25H36N2O4/c28-24(30-19-21-11-4-1-5-12-21)17-10-18-25(29)31-27(23-15-8-3-9-16-23)20-26-22-13-6-2-7-14-22/h1,4-5,11-12,20,22-23H,2-3,6-10,13-19H2/b26-20+
LEDER O=C(CCCC(=O)ON(\C=N\C1CCCCC1)C2CCCCC2)OCc3ccccc3
IUPAC-namn 1-O-benzyl 5-O-[cyclohexyl-[(E)-cyclohexyliminomethyl]amino] pentanedioate
Molekylvikt (g/mol) 428.56
Formel vikt 428.2675