Learn More
Alfa Aesar™ 1-Benzyl-6-cyanoindole, 97%
Brand:  Alfa Aesar™ H63525.03
Additional Details : CAS Number : 1030423-43-8 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity 1g
 View more versions of this product
Product Code. 15470259
  • 1166.62 SEK / 1g
Estimated Shipment date
from Supplier 15-08-2018
Add to basket
Specifications
Chemical Identifiers
1-Benzyl-6-cyanoindole
MFCD15200961
1-Benzylindole-6-carbonitrile
1030423-43-8
UN3439
Specifications
1-Benzyl-6-cyanoindole
1030423-43-8
Solid
1g
MFCD15200961
1-Benzylindole-6-carbonitrile
C1=CC=C(C=C1)CN2C=CC3=C2C=C(C=C3)C#N
232.286
232.28
Off-white to brown
97%
C16H12N2
UN3439
UYUKKLGDRIKDEO-UHFFFAOYSA-N
1-benzylindole-6-carbonitrile
29012020