Läs mer
1-(Benzyloxy)-4-bromo-2-methoxybenzene, TRC
Produktkod. 30331710
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30331710 100 mg
30342530 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30331710 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB291493100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(Benzyloxy)-4-bromo-2-methoxybenzene
CAS 63057-72-7
Molekylformel C14H13BrO2
Kvantitet 100 mg
InChI formel InChI=1S/C14H13BrO2/c1-16-14-9-12(15)7-8-13(14)17-10-11-5-3-2-4-6-11/h2-9H,10H2,1H3
LEDER COc1cc(Br)ccc1OCc2ccccc2
IUPAC-namn 4-bromo-2-methoxy-1-phenylmethoxybenzene
Molekylvikt (g/mol) 293.16
Formel vikt 292.0099