Läs mer
1-BOC-2,3-Dihydropyrrole-4-boronic acid, pinacol ester, TRC
Produktkod. 30340703
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30340703 25 mg
30342628 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30340703 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB20187825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-BOC-2,3-Dihydropyrrole-4-boronic acid, pinacol ester
CAS 1401165-14-7
Molekylformel C15 H26 B N O4
Kvantitet 25 mg
Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-pyrrole-1-carboxylic Acid tert-Butyl Ester
InChI formel InChI=1S/C15H26BNO4/c1-13(2,3)19-12(18)17-9-8-11(10-17)16-20-14(4,5)15(6,7)21-16/h10H,8-9H2,1-7H3
LEDER CC(C)(C)OC(=O)N1CCC(=C1)B2OC(C)(C)C(C)(C)O2
IUPAC-namn tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydropyrrole-1-carboxylate
Molekylvikt (g/mol) 295.182
Formel vikt 295.195