Learn More
1-Boc-2-phenylpiperazine, 97%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Description
1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Spécification
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 262.35 |
Percent Purity | 97% |
Chemical Name or Material | 1-Boc-2-phenylpiperazine |