missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-(methylamino)azetidine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents.
1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents.
Specifications
Solubility Information | Slightly soluble in water. |
Formula Weight | 186.25 |
Percent Purity | 95% |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Chemical Name or Material | 1-Boc-3-(methylamino)azetidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'