missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(4-formylphenyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog Number p-7036934
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
197638-83-8
C16H22N2O3
290.36
MFCD05864663
KHORERZDMJTBMR-UHFFFAOYSA-N
1-boc-4-4-formylphenyl piperazine, tert-butyl 4-4-formylphenyl piperazine-1-carboxylate, 4-4-tert-butoxycarbonyl piperazinebenzaldehyde, tert-butyl 4-4-formylphenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-formylphenyl piperazinecarboxylate, acmc-209f1h, 1-n-boc-4-4-formylphenyl piperazine, 4-4-tert-butoxycarbonylpiperazin-1-yl benzaldehyde
2795509
tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
197638-83-8
C16H22N2O3
290.36
MFCD05864663
KHORERZDMJTBMR-UHFFFAOYSA-N
1-boc-4-4-formylphenyl piperazine, tert-butyl 4-4-formylphenyl piperazine-1-carboxylate, 4-4-tert-butoxycarbonyl piperazinebenzaldehyde, tert-butyl 4-4-formylphenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-formylphenyl piperazinecarboxylate, acmc-209f1h, 1-n-boc-4-4-formylphenyl piperazine, 4-4-tert-butoxycarbonylpiperazin-1-yl benzaldehyde
2795509
tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 118°C to 121°C
Quantity 1 g
UN Number UN2811
Sensitivity Air Sensitive
Formula Weight 290.36
Percent Purity 97%
Chemical Name or Material 1-Boc-4-(4-formylphenyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'