missing translation for 'onlineSavingsMsg'
Läs mer

1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid, 96%

Produktkod. p-7033076
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11414510 250 mg 250mg
11424510 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11414510 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H50086.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1210863-93-6 | C12H19NO6 | 273.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1210863-93-6
Molekylformel C12H19NO6
Molekylvikt (g/mol) 273.29
MDL-nummer MFCD07781134
InChI-nyckel OHQXZMNJROMWMW-UHFFFAOYNA-N
Synonym 4-acetoxy-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, 1-boc-4-methoxycarbonyl pyrrolidine-3-carboxylic acid, pyrrolidine-1,3,4-tricarboxylic acid 1-tert-butyl 3-methyl diester, 4-acetyloxy-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid
PubChem CID 45925717
IUPAC-namn 4-acetyloxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
LEDER CC(=O)OC1CN(CC1C(=O)O)C(=O)OC(C)(C)C

Specifications

Kemiskt namn eller material 1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid
Kvantitet 250 mg
Formel vikt 273.29
Procent renhet 96%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.