Läs mer
1-Bromo-1-cyclopentyl 4-Chlorophenyl Ketone, TRC
Produktkod. 30335729
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
100 mg
Produktkod. Kvantitet
30335729 100 mg
1 missing translation for 'options'
Produktkod. 30335729 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB682885100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Bromo-1-cyclopentyl 4-Chlorophenyl Ketone
CAS 6728-56-9
Molekylformel C12H12BrClO
Kvantitet 100 mg
Synonym 1-Bromocyclopentyl p-Chlorophenyl Ketone, (1-Bromocyclopentyl)(4-chlorophenyl)methanone
InChI formel InChI=1S/C12H12BrClO/c13-12(7-1-2-8-12)11(15)9-3-5-10(14)6-4-9/h3-6H,1-2,7-8H2
LEDER Clc1ccc(cc1)C(=O)C2(Br)CCCC2
IUPAC-namn (1-bromocyclopentyl)-(4-chlorophenyl)methanone
Molekylvikt (g/mol) 287.58
Formel vikt 285.976