Läs mer
1-Bromo-2-(2-propen-1-yloxy)-benzene, TRC
Produktkod. 30337092
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30337092 25 g
1 options
Produktkod. 30337092 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68760025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Bromo-2-(2-propen-1-yloxy)-benzene
CAS 60333-75-7
Molekylformel C9H9BrO
Kvantitet 25 g
Synonym 1-Bromo-2-(2-propenyloxy)-benzene, 1-(Allyloxy)-2-bromobenzene, 1-Bromo-2-allyloxybenzene, Allyl 2-bromophenyl Ether, Allyl o-Bromophenyl Ether, o-(Allyloxy)bromobenzene, o-Bromophenyl Allyl Ether
InChI formel InChI=1S/C9H9BrO/c1-2-7-11-9-6-4-3-5-8(9)10/h2-6H,1,7H2
LEDER Brc1ccccc1OCC=C
IUPAC-namn 1-bromo-2-prop-2-enoxybenzene
Molekylvikt (g/mol) 213.07
Formel vikt 211.9837