Läs mer
1-Bromo-2-phenoxybenzene, TRC
Produktkod. 30180322
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180322 1 g
30179908 100 mg
30180433 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180322 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6872681G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7025-06-1
Molekylformel C12 H9 Br O
Molekylvikt (g/mol) 249.1
Synonym Benzene, 1-bromo-2-phenoxy-, Ether, o-bromophenyl phenyl (6CI,7CI,8CI), 1-Bromo-2-phenoxybenzene, 2-Bromodiphenyl ether, 2-Bromophenyl phenyl ether, 2-Phenoxy-1-bromobenzene, 2-Phenoxybromobenzene, BDE 1, PBDE 1, o-Bromophenyl phenyl ether
IUPAC-namn 1-bromo-2-phenoxybenzene
LEDER Brc1ccccc1Oc2ccccc2
Specifikationer
Kemiskt namn eller material PBDE 1 (2-Bromodiphenyl Ether)
Kvantitet 1 g
InChI formel InChI=1S/C12H9BrO/c13-11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9H
Formel vikt 247.9837