Learn More
1-Bromo-2-propanol, 70%, remainder 2-bromo-1-propanol, tech., ACROS Organics™
Brand:  Acros Organics 218620500
Additional Details : CAS Number : 19686-73-8 Weight : 0.12650kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Packaging Glass bottle
Quantity 50mL
 View more versions of this product
Product Code. 10006143
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Specifications
Chemical Identifiers
19686-73-8
138.992
WEGOLYBUWCMMMY-UHFFFAOYSA-N
C3H7BrO
MFCD00004529
29740
Specifications
1-Bromo-2-propanol
65.0
100.0
1.4790 to 1.4900
145.0°C to 148.0°C
50mL
70%
70%
CH3CH(OH)CH2Br
01, 365
CC(CBr)O
138.992
138.99
598-18-5
Authentic
Glass bottle
1.5300g/mL
Technical Grade
54°C
Colorless through Brown
C3H7BrO
MFCD00004529
WEGOLYBUWCMMMY-UHFFFAOYSA-N
1-bromopropan-2-ol
29740
Liquid
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 1-Bromo-2-propanol, remainder 2-bromo-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
SDS