Läs mer
1-Bromo-3-(4-bromophenoxy)benzene, TRC
Produktkod. 30336651
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30336651 25 mg
30334051 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30336651 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB67999225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material PBDE 13 (3,4'-Dibromodiphenyl Ether)
CAS 83694-71-7
Molekylformel C12 H8 Br2 O
Kvantitet 25 mg
Synonym Benzene, 1-bromo-3-(4-bromophenoxy)-, 1-Bromo-3-(4-bromophenoxy)benzene, 3,4'-Dibromodiphenyl ether, BDE 13, PBDE 13
InChI formel InChI=1S/C12H8Br2O/c13-9-4-6-11(7-5-9)15-12-3-1-2-10(14)8-12/h1-8H
LEDER Brc1ccc(Oc2cccc(Br)c2)cc1
IUPAC-namn 1-bromo-3-(4-bromophenoxy)benzene
Molekylvikt (g/mol) 328
Formel vikt 325.8942