missing translation for 'onlineSavingsMsg'
Läs mer

1-Bromo-3,5,7-trimethyladamantane, TRC

Produktkod. 30345351
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30345351 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345351 Supplier Toronto Research Chemicals Supplier No. TRCB6839451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-Bromo-3,5,7-trimethyladamantane
CAS 53398-55-3
Molekylformel C13 H21 Br
Kvantitet 1 g
Synonym 1-Bromo-3,5,7-trimethyl-tricyclo[3.3.1.13,7]decane, NSC 251758
InChI formel InChI=1S/C13H21Br/c1-10-4-11(2)6-12(3,5-10)9-13(14,7-10)8-11/h4-9H2,1-3H3/t10-,11+,12-,13-
LEDER C[C@@]12C[C@@]3(C)C[C@@](C)(C1)C[C@](Br)(C3)C2
IUPAC-namn 1-bromo-3,5,7-trimethyladamantane
Molekylvikt (g/mol) 257.21
Formel vikt 256.0827
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.