missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | -16°C |
Density | 1.698 |
Boiling Point | 152°C to 155°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.427 |
Quantity | 10 g |
Beilstein | 2123669 |
Solubility Information | Immiscible with water. |
Formula Weight | 293.01 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'