Learn More
1-Bromo-3,5-dimethyladamantane, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 941-37-7 |
| Molecular Formula | C12H19Br |
| MDL Number | MFCD00077197 |
| Quantity | 5 g |
| SMILES | CC1(C2)CC3(C)CC2(Br)CC(C3)C1 |
| Molecular Weight (g/mol) | 243.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Bromo-3,5-dimethyladamantane |