missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-5-fluorobenzene, 98%, Thermo Scientific™

Catalog Number p-4812599
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
33863-76-2
C6H3BrClF
209.45
GGMDFPMASIXEIR-UHFFFAOYSA-N
3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene
2736223
1-bromo-3-chloro-5-fluorobenzene
C1=C(C=C(C=C1Cl)Br)F
This item is not returnable. View return policy
33863-76-2
C6H3BrClF
209.45
GGMDFPMASIXEIR-UHFFFAOYSA-N
3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene
2736223
1-bromo-3-chloro-5-fluorobenzene
C1=C(C=C(C=C1Cl)Br)F
Density 1.7200g/mL
Boiling Point 72°C (5.0 mmHg)
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)F
Quantity 5 g
Specific Gravity 1.72
Formula Weight 209.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-chloro-5-fluorobenzene
compliance-icons
Product Identifier
  • 1-Bromo-3-chloro-5-fluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'