missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Bromomethyl)-3-(trifluoromethoxy)benzene, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 97.0 |
Density | 1.5720g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | CH2BrC6H4OCF3 |
Quantity | 1 g |
Formula Weight | 255.03 |
Percent Purity | 97% |
Chemical Name or Material | 1-(Bromomethyl)-3-(trifluoromethoxy)benzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'