Learn More
1-Bromononane, TRC
Product Code. 30343967
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30343967 5 g 5g
1 options
Product Code. 30343967 Supplier Toronto Research Chemicals Supplier No. TRCB6875755G
 Restricted Item
Beräknad leverans: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromononane
CAS 693-58-3
Molecular Formula C9H19Br
Quantity 5 g
Synonym 1-Bromo-n-nonane, 1-Nonyl Bromide, 1-n-Nonyl bromide, NSC 5519, Nonyl Bromide, n-Nonyl bromide
InChI Formula InChI=1S/C9H19Br/c1-2-3-4-5-6-7-8-9-10/h2-9H2,1H3
SMILES CCCCCCCCCBr
IUPAC Name 1-bromononane
Molecular Weight (g/mol) 207.15
Formula Weight 206.067