missing translation for 'onlineSavingsMsg'
Läs mer

DSG1 Rabbit anti-Human, Polyclonal, Proteintech

Produktkod. 16843075 Handla allt Proteintech Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
20 μL
150 μL
Förpackningsstorlek:
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16843075 20 μL 20µL
16833075 150 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16843075 Leverantör Proteintech Leverantörsnummer 245871AP20UL

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.

Specifikationer

Antigen DSG1
Användningsområden Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Klassificering Polyclonal
Koncentration 0.59 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gen DSG1
Genaccessionsnr. Q02413
Gene Alias Cadherin family member 4, CDHF4, desmoglein 1, Desmosomal glycoprotein 1, DG1, DGI, DSG, DSG1, Pemphigus foliaceus antigen, PPKS1, SPPK1
Gensymboler DSG1
Värd art Rabbit
Immunogen DSG1 Fusion Protein Ag20184
Reningsmetod Antigen Affinity Chromatography
Kvantitet 20 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 1828
Målarter Human
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.