Läs mer
1-C-[3-[[5-(4-Fluorophenyl)-2-thienyl]methyl]-4-methylphenyl]-D-glucose, TRC
Produktkod. 30179203
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179203 50 mg
30179545 250 mg
30179651 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179203 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF82821050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1809403-04-0
Molekylformel C24 H25 F O6 S
Molekylvikt (g/mol) 460.52
Synonym (2R,3S,4R,5R)-1-(3-((5-(4-Fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)-2,3,4,5,6-pentahydroxyhexan-1-one
IUPAC-namn (2R,3S,4R,5R)-1-[3-[[5-(4-fluorophenyl)thiophen-2-yl]methyl]-4-methylphenyl]-2,3,4,5,6-pentahydroxyhexan-1-one
LEDER Cc1ccc(cc1Cc2ccc(s2)c3ccc(F)cc3)C(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
Specifikationer
Kemiskt namn eller material 1-C-(3-((5-(4-Fluorophenyl)-2-thienyl)methyl)-4-methylphenyl)-D-glucose
Kvantitet 50 mg
InChI formel InChI=1S/C24H25FO6S/c1-13-2-3-15(21(28)23(30)24(31)22(29)19(27)12-26)10-16(13)11-18-8-9-20(32-18)14-4-6-17(25)7-5-14/h2-10,19,22-24,26-27,29-31H,11-12H2,1H3/t19-,22-,23+,24+/m1/s1
Formel vikt 460.1356