missing translation for 'onlineSavingsMsg'
Läs mer

1-(Karboxymetyl)cyklohexankarboxylsyra, Mikromol™

Product Code. 30049929
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30049929 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049929 Supplier Mikromol Supplier No. MM0684.070025
 Begränsad artikel
Beräknad leverans: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol – din pålitliga källa för högkvalitativa läkemedelsreferensstandarder. Stöder noggrann, regelrätt analys med material som är ackrediterat 17034 ISO och utformat för att säkerställa förtroende för varje resultat.

1-(Karboxymetyl)cyklohexankarboxylsyra
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-(Carboxymethyl)cyclohexanecarboxylic Acid
CAS 67950-95-2
Analyt- eller komponentnamn 1-(Carboxymethyl)cyclohexanecarboxylic Acid
Frakt skick Rumstemperatur
Molekylformel C9 H14 O4
Certifikat och efterlevnad ISO 9001
Synonym Gabapentin Imp. E (EP), Gabapentin USP Related Compound E, Gabapentin USP RC E, 1-(Carboxymethyl)cyclohexanecarboxylic Acid, Gabapentin Related Compound E, Gabapentin Impurity E
InChI formel InChI=1S/C9H14O4/c10-7(11)6-9(8(12)13)4-2-1-3-5-9/h1-6H2,(H,10,11)(H,12,13)
LEDER OC(=O)CC1(CCCCC1)C(=O)O
IUPAC-namn 1-(karboxymetyl)cyklohexan-1-karboxylsyra
Molekylvikt (g/mol) 186.21
Kvantitet 25 mg
Formel vikt 186,0892 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.