Läs mer
1-Chloro-2-propanol Phosphorochloridate, TRC
Produktkod. 30365046
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30365046 25 mg
1 options
Produktkod. 30365046 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38022525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Chloro-2-propanol Phosphorochloridate
CAS 13674-83-4
Molekylformel C6H12Cl3O3P
Kvantitet 25 mg
Synonym Phosphorochloridic Acid, Bis(2-chloro-1-methylethyl) Ester
InChI formel InChI=1S/C6H12Cl3O3P/c1-5(3-7)11-13(9,10)12-6(2)4-8/h5-6H,3-4H2,1-2H3
LEDER CC(CCl)OP(=O)(Cl)OC(C)CCl
IUPAC-namn 1-chloro-2-[chloro(1-chloropropan-2-yloxy)phosphoryl]oxypropane
Molekylvikt (g/mol) 269.49
Formel vikt 267.959