Läs mer
1-Chloro-4-methoxybutane, TRC
Produktkod. 30349459
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30349459 500 mg
1 options
Produktkod. 30349459 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC367940500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Chloro-4-methoxybutane
CAS 17913-18-7
Molekylformel C5H11ClO
Kvantitet 500 mg
Synonym 1-Chloro-4-methoxybutane, 4-Chlorobutyl Methyl Ether, 4-Methoxy-1-chlorobutane, 4-Methoxybutyl Chloride, Methyl 4-chlorobutyl Ether, NSC 83550, delta-Chlorobutyl Methyl Ether
InChI formel InChI=1S/C5H11ClO/c1-7-5-3-2-4-6/h2-5H2,1H3
LEDER COCCCCCl
IUPAC-namn 1-chloro-4-methoxybutane
Molekylvikt (g/mol) 122.59
Formel vikt 122.0498