Läs mer
1-Chloroethyl Cyclohexyl Carbonate, TRC
Produktkod. 30004341
Change view
Kvantitet:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004341 100 g
30011603 10 g
30012490 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004341 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC367640100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99464-83-2
Molekylformel C9 H15 Cl O3
Molekylvikt (g/mol) 206.67
Synonym Carbonic acid, 1-chloroethyl cyclohexyl ester, 1-(Cyclohexyloxycarbonyloxy)ethyl chloride, 1-Chloroethyl cyclohexyl carbonate, 1-[[(Cyclohexyloxy)carbonyl]oxy]-1-chloroethane, Carbonic acid 1-chloroethyl ester cyclohexyl ester, Cilexetil chloride, Cyclohexyl 1-chloroethyl carbonate, Cyclohexyl 1-Chloroethyl Carbonate
IUPAC-namn 1-chloroethyl cyclohexyl carbonate
LEDER CC(Cl)OC(=O)OC1CCCCC1
Specifikationer
Kemiskt namn eller material Cyclohexyl 1-Chloroethyl Carbonate
Kvantitet 100 g
InChI formel InChI=1S/C9H15ClO3/c1-7(10)12-9(11)13-8-5-3-2-4-6-8/h7-8H,2-6H2,1H3
Formel vikt 206.071