missing translation for 'onlineSavingsMsg'
Läs mer

1-Chloroethyl Cyclohexyl Carbonate, TRC

Produktkod. 30012490
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012490 1 g
30004341 100 g
30011603 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30012490 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3676401G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99464-83-2
Molekylformel C9 H15 Cl O3
Molekylvikt (g/mol) 206.67
Synonym Carbonic acid, 1-chloroethyl cyclohexyl ester, 1-(Cyclohexyloxycarbonyloxy)ethyl chloride, 1-Chloroethyl cyclohexyl carbonate, 1-[[(Cyclohexyloxy)carbonyl]oxy]-1-chloroethane, Carbonic acid 1-chloroethyl ester cyclohexyl ester, Cilexetil chloride, Cyclohexyl 1-chloroethyl carbonate, Cyclohexyl 1-Chloroethyl Carbonate
IUPAC-namn 1-chloroethyl cyclohexyl carbonate
LEDER CC(Cl)OC(=O)OC1CCCCC1

Specifications

Kemiskt namn eller material Cyclohexyl 1-Chloroethyl Carbonate
Kvantitet 1 g
InChI formel InChI=1S/C9H15ClO3/c1-7(10)12-9(11)13-8-5-3-2-4-6-8/h7-8H,2-6H2,1H3
Formel vikt 206.071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.