missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorophthalazine, 97%, Thermo Scientific Chemicals
451.28 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
10 g
Description
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.
Solubility
Insoluble in water. Soluble in dichloromethane, ethanol and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water, heat, moisture sensitive.
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.
Solubility
Insoluble in water. Soluble in dichloromethane, ethanol and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water, heat, moisture sensitive.
Specifications
Melting Point | 109°C to 112°C |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. Soluble in dichloromethane,ethanol and methanol. |
Formula Weight | 164.59 |
Percent Purity | 97% |
Chemical Name or Material | 1-Chlorophthalazine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'