missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.
Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with oxidizing agents, Strong acids, Strong bases, Strong reducing agents, Acid chlorides, Acid anhydrides.
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.
Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with oxidizing agents, Strong acids, Strong bases, Strong reducing agents, Acid chlorides, Acid anhydrides.
Specifications
Melting Point | 118°C to 122°C |
Quantity | 5 g |
Beilstein | 6286 |
Sensitivity | Hygroscopic |
Formula Weight | 163.18 |
Percent Purity | 97% |
Chemical Name or Material | 1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'