Learn More
Alfa Aesar™ 1-Cyclohexenyl trifluoromethanesulfonate, 97%
Brand:  Alfa Aesar™ H55879.06
Additional Details : CAS Number : 28075-50-5 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15472268
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
View alternative products »
Description
Description
Applications
Used as a pharmaceutical intermediate.
Notes
Store at 4°C.
Specifications
Chemical Identifiers
1-Cyclohexenyl trifluoromethanesulfonate
28075-50-5
MFCD06411281
Specifications
1-Cyclohexenyl trifluoromethanesulfonate
28075-50-5
97%
MFCD06411281
C1CCC(=CC1)OS(=O)(=O)C(F)(F)F
319440
84° to 87°C (4mmHg)
5g
230.2
Liquid
C7H9F3O3S
WVSCRRLWRRANJY-UHFFFAOYSA-N
230.201
1.315g/mL
Colorless to yellow