Läs mer
1-Cyclohexyl-3-[[4-[2-[[(6-methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]urea, Mikromol™
Produktkod. 30049645
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049645 25 mg
30052532 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049645 Leverantör Mikromol Leverantörsnummer MM0942.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Cyclohexyl-3-[[4-[2-[[(6-methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]urea
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Cyclohexyl-3-[(4-[2-[((6-methylpyrazin-2-yl)carbonyl)amino]ethyl]phenyl)sulphonyl]urea
CAS 66375-96-0
Analyt- eller komponentnamn 1-Cyclohexyl-3-[(4-[2-[((6-methylpyrazin-2-yl)carbonyl)amino]ethyl]phenyl)sulphonyl]urea
Frakt skick Room Temperature
Molekylformel C21 H27 N5 O4 S
Certifikat och efterlevnad ISO 9001
Synonym Glipizide Imp. E (EP), Glipizide USP Related Compound C, Glipizide USP RC C, 1-Cyclohexyl-3-[[4-[2-[[(6-methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]urea, Glipizide Related Compound C, Glipizide Impurity E
InChI formel InChI=1S/C21H27N5O4S/c1-15-13-22-14-19(24-15)20(27)23-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17H,2-6,11-12H2,1H3,(H,23,27)(H2,25,26,28)
LEDER Cc1cncc(n1)C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3
IUPAC-namn N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-6-methylpyrazine-2-carboxamide
Molekylvikt (g/mol) 445.54
Kvantitet 25 mg
Formel vikt 445.1784 g/mol