Läs mer
1-Cyclohexylnaphthalene, TRC
Produktkod. 30349756
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349756 10 g
30366897 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30349756 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98805510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3042-69-1
Molekylformel C16H18
Molekylvikt (g/mol) 210.31
Synonym 1-Cyclohexyl-naphthalene, NSC 131573
IUPAC-namn 1-cyclohexylnaphthalene
LEDER C1CCC(CC1)C2=CC=CC3=CC=CC=C32
Specifikationer
Kemiskt namn eller material 1-Cyclohexylnaphthalene
Kvantitet 10 g
InChI formel InChI=1S/C16H18/c1-2-7-13(8-3-1)16-12-6-10-14-9-4-5-11-15(14)16/h4-6,9-13H,1-3,7-8H2
Formel vikt 210.1409