missing translation for 'onlineSavingsMsg'
Läs mer

1-Cyclopropyl-2-(2-fluorophenyl)-2-hydroxy-ethanone, TRC

Produktkod. 30354712
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354712 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30354712 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9894651G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-Cyclopropyl-2-(2-fluorophenyl)-2-hydroxy-ethanone
CAS 1100905-45-0
Molekylformel C11 H11 F O2
Kvantitet 1 g
Synonym Prasugrel alpha-Hydroxy Impurity
InChI formel InChI=1S/C11H11FO2/c12-9-4-2-1-3-8(9)11(14)10(13)7-5-6-7/h1-4,7,11,14H,5-6H2
LEDER OC(C(=O)C1CC1)c2ccccc2F
IUPAC-namn 1-cyclopropyl-2-(2-fluorophenyl)-2-hydroxyethanone
Molekylvikt (g/mol) 194.202
Formel vikt 194.074
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.